Di-fmoc-D-α,β-diaminopropionic acid 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Dap(Fmoc)-OH, (R)-2,3-Bis((((9H-fluoren-9-yl)methoxy)carbonyl)amino)propanoic acid

A916688-1G A916688-250MG
77019-950EA 158.08 USD
77019-950 77019-952
Di-fmoc-D-α,β-diaminopropionic acid 95%
Di-fmoc-D-alpha,beta-diaminopropionic acid

Precaución: for Research Use Only

Formula: C₃₃H₂₈N₂O₆
MW: 548.6 g/mol
MDL Number: MFCD01632030
CAS Number: 1217631-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR