(S)-2-Bromobutanoic acid 98%

Supplier: Ambeed
Danger

A914267-25G A914267-1G A914267-5G
77007-076EA 255 USD
77007-076 77573-836 77603-284
(S)-2-Bromobutanoic acid 98%
(S)-2-Bromobutanoic acid

Precaución: for Research Use Only

Formula: C₄H₇BrO₂
MDL Number: MFCD00210112
CAS Number: 32659-49-7
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR