Bis(2-butoxyethyl)ether 98%
Supplier: Ambeed
|
Warning
|
A914058-1000G
A914058-100G
A914058-25G
A914058-500G
77007-068EA
25
USD
77007-068
77007-070
77007-072
77007-074
Bis(2-butoxyethyl)ether 98%
Bis(2-butoxyethyl)ether
Precaución: for Research Use Only
Formula:
C₁₂H₂₆O₃ |
MDL Number:
MFCD00009459 CAS Number: 112-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...