5-Amino-1,2,4-thiadiazole 98%

Supplier: Ambeed
Warning

Synonyms: 1,2,4-Thiadiazol-5-amine

A913572-10G A913572-1G A913572-250MG A913572-25G A913572-5G
77007-058EA 77 USD
77007-058 77007-060 77007-062 77007-064 77007-066
5-Amino-1,2,4-thiadiazole 98%
5-Amino-1,2,4-thiadiazole

Precaución: for Research Use Only

Formula: C₂H₃N₃S
MDL Number: MFCD01102171
CAS Number: 7552-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR