7-(Hydroxymethyl)-1H-indazole 95%

Supplier: Ambeed
Warning

Synonyms: 1H-Indazol-7-ylmethanol, 7-(Hydroxymethyl)indazole

A911971-1G A911971-250MG A911971-5G
77036-870EA 58.35 USD
77036-870 77036-872 77036-874
7-(Hydroxymethyl)-1H-indazole 95%
7-(Hydroxymethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.162 g/mol
MDL Number: MFCD11052637
CAS Number: 1092961-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR