8-Bromoimidazo[1,2-a]pyrazine 97%

Supplier: Ambeed
Warning

A911737-1G A911737-250MG A911737-5G
77036-864EA 166.57 USD
77036-864 77036-866 77036-868
8-Bromoimidazo[1,2-a]pyrazine 97%
8-Bromoimidazo[1,2-a]pyrazine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.02 g/mol
MDL Number: MFCD18072715
CAS Number: 69214-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR