4-Iodobenzamidine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 4-Iodobenzene-1-carboximidamide hydrochloride, 4-Iodobenzimidamide hydrochloride, 4-Iodophenylamidine hydrochloride

A911535-100MG A911535-1G A911535-250MG
77019-944EA 48 USD
77019-944 77019-946 77019-948
4-Iodobenzamidine hydrochloride 98%
4-Iodobenzamidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈ClIN₂
MW: 282.51
MDL Number: MFCD04114465
CAS Number: 134322-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR