trans-4-(Aminomethyl)cyclohexyl)methanol 95%

Supplier: Ambeed
Danger

Synonyms: trans-4-(Aminomethyl)cyclohexylmethanol

A911187-100MG A911187-1G A911187-250MG A911187-5G
77019-938EA 16 USD
77019-938 77019-940 77019-942 77891-681
trans-4-(Aminomethyl)cyclohexyl)methanol 95%
trans-4-(Aminomethyl)cyclohexyl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₇NO
MDL Number: MFCD14702919
CAS Number: 17879-23-1
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR