3-Bromo-9-(p-tolyl)-9H-carbazole 97%

Supplier: Ambeed
Warning

A906977-1G A906977-250MG A906977-5G
76993-854EA 29 USD
76993-854 76993-856 76993-858
3-Bromo-9-(p-tolyl)-9H-carbazole 97%
3-Bromo-9-(p-tolyl)-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₉H₁₄BrN
MDL Number: MFCD16658912
CAS Number: 731016-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR