2-Chloro-4-methoxybenzoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Chloro-p-anisic acid
A906644-100G
A906644-10G
A906644-1G
A906644-250MG
A906644-25G
A906644-5G
77007-040EA
516.66
USD
77007-040
77007-042
77007-044
77007-046
77007-048
77007-050
2-Chloro-4-methoxybenzoic acid 98%
2-Chloro-4-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇ClO₃ |
MDL Number:
MFCD00085943 CAS Number: 21971-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...