4-Fluoro-2,6-dimethylphenol 97%

Supplier: Ambeed
Warning

A906083-1G A906083-250MG
76993-850EA 72.15 USD
76993-850 76993-852
4-Fluoro-2,6-dimethylphenol 97%
4-Fluoro-2,6-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.16 g/mol
MDL Number: MFCD04972873
CAS Number: 2338-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR