Ethyl 2-(2-fluorobenzyl)-3-oxobutanoate 95%
Supplier: Ambeed
|
Warning
|
A905999-100MG
A905999-1G
A905999-250MG
A905999-25G
A905999-5G
77007-030EA
11.67
USD
77007-030
77007-032
77007-034
77007-036
77007-038
Ethyl 2-(2-fluorobenzyl)-3-oxobutanoate 95%
Ethyl 2-(2-fluorobenzyl)-3-oxobutanoate
Precaución: for Research Use Only
Formula:
C₁₃H₁₅FO₃ MW: 238.26 g/mol |
MDL Number:
MFCD03093821 CAS Number: 24106-86-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...