(R)-1-(2,4-Dichlorophenyl)ethanamine 95%
Supplier: Ambeed
|
Warning
|
A905805-100MG
A905805-250MG
A905805-5G
A905805-1G
76993-844EA
40.32
USD
76993-844
76993-846
76993-848
77684-863
(R)-1-(2,4-Dichlorophenyl)ethanamine 95%
(R)-1-(2,4-Dichlorophenyl)ethanamine
Precaución: For research use only.
Formula:
C₈H₉Cl₂N MW: 190.07 g/mol |
MDL Number:
MFCD06761788 CAS Number: 133773-29-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...