5-Bromo-4-fluoro-2-iodotoluene 98%

Supplier: Ambeed
Warning

A905753-1G A905753-250MG A905753-5G
76993-838EA 26.53 USD
76993-838 76993-840 76993-842
5-Bromo-4-fluoro-2-iodotoluene 98%
5-Bromo-4-fluoro-2-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFI
MDL Number: MFCD09800686
CAS Number: 861928-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR