1-Ethyl-1H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-Ethylpyrazole-3-carboxylic Acid

A905376-250MG A905376-1G
77036-860EA 23.34 USD
77036-860 77603-262
1-Ethyl-1H-pyrazole-3-carboxylic acid 95%
1-Ethyl-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD02090832
CAS Number: 400755-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR