3-Hydroxy-3-phenylpropionitrile 97%

Supplier: Ambeed
Danger

A905033-250MG A905033-100MG A905033-5G
77019-936EA 44 USD
77019-936 77891-676 77891-677
3-Hydroxy-3-phenylpropionitrile 97%
3-Hydroxy-3-phenylpropionitrile

Precaución: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD00544189
CAS Number: 17190-29-3
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR