2-[(Dimethylamino)methyl]benzoic Acid 95%

Supplier: Ambeed
Warning

A903783-100MG A903783-250MG
77019-930EA 32 USD
77019-930 77019-932
2-[(Dimethylamino)methyl]benzoic Acid 95%
2-[(Dimethylamino)methyl]benzoic Acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD11841286
CAS Number: 55741-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR