(6-((4-Methoxybenzyl)carbamoyl)pyridin-3-yl)boronic acid 96%

Supplier: Ambeed
Warning

A903459-100MG A903459-1G A903459-250MG A903459-5G
77019-922EA 39 USD
77019-922 77019-924 77019-926 77019-928
(6-((4-Methoxybenzyl)carbamoyl)pyridin-3-yl)boronic acid 96%
(6-((4-Methoxybenzyl)carbamoyl)pyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₅BN₂O₄
MDL Number: MFCD10699628
CAS Number: 1072946-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR