5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 97%
Supplier: Ambeed
|
Warning
|
A902939-100MG
A902939-1G
76993-828EA
46.68
USD
76993-828
76993-830
5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde 97%
5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde
Precaución: for Research Use Only
Formula:
C₈H₅ClN₂O |
MDL Number:
MFCD12962789 CAS Number: 954112-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...