4,5,9,10-Tetrabromo-2,7-bis(2-octyldodecyl)benzo[lmn][3,8]phenanthroline-1,3,6,8(2H,7H)-tetraone 98%

Supplier: Ambeed
Warning

A902665-100MG A902665-250MG
77019-918EA 140.04 USD
77019-918 77019-920
4,5,9,10-Tetrabromo-2,7-bis(2-octyldodecyl)benzo[lmn][3,8]phenanthroline-1,3,6,8(2H,7H)-tetraone 98%
4,5,9,10-Tetrabromo-2,7-bis(2-octyldodecyl)benzo[lmn][3,8]phenanthroline-1,3,6,8(2H,7H)-tetraone

Precaución: for Research Use Only

Formula: C₅₄H₈₂Br₄N₂O₄
MW: 1142.85 g/mol
MDL Number: MFCD27923040
CAS Number: 1219501-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR