3-Amino-N-butylbenzamide 98%

Supplier: Ambeed
Warning

A901555-10G A901555-1G A901555-5G
77007-020EA 366.02 USD
77007-020 77007-022 77007-024
3-Amino-N-butylbenzamide 98%
3-Amino-N-butylbenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₆N₂O
MDL Number: MFCD00088668
CAS Number: 6837-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR