2-Chloro-1-(2,7-dichloro-9H-fluoren-4-yl)ethanone 95%

Supplier: Ambeed
Warning

A901113-250MG A901113-5G
77019-906EA 15.92 USD
77019-906 77019-908
2-Chloro-1-(2,7-dichloro-9H-fluoren-4-yl)ethanone 95%
2-Chloro-1-(2,7-dichloro-9H-fluoren-4-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₅H₉Cl₃O
MW: 311.59 g/mol
MDL Number: MFCD11846253
CAS Number: 131023-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR