N-(3-(Trimethoxysilyl)propyl)ethylenediamine 97%

Supplier: Ambeed
Danger

A900254-25G A900254-100G
77007-014EA 10 USD
77007-014 77603-246
N-(3-(Trimethoxysilyl)propyl)ethylenediamine 97%
N-(3-(Trimethoxysilyl)propyl)ethylenediamine

Precaución: for Research Use Only

Formula: H₂NCH₂CH₂NHCH₂CH₂CH₂Si(OCH₃)₃
MDL Number: MFCD00008173
CAS Number: 1760-24-3
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR