Methyl-3-(cyclopentylamino)propanoate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-(cyclopentylamino)propanoate

A899115-10G A899115-1G A899115-250MG A899115-5G
77007-006EA 463.62 USD
77007-006 77007-008 77007-010 77007-012
Methyl-3-(cyclopentylamino)propanoate 95%
Methyl-3-(cyclopentylamino)propanoate

Precaución: for Research Use Only

Formula: C₉H₁₇NO₂
MW: 171.2368 g/mol
MDL Number: MFCD10687312
CAS Number: 754125-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR