5-Bromo-2-(methylamino)benzoic acid 95%

Supplier: Ambeed
Warning

A898562-1G A898562-250MG
77019-896EA 51 USD
77019-896 77019-898
5-Bromo-2-(methylamino)benzoic acid 95%
5-Bromo-2-(methylamino)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MDL Number: MFCD07439747
CAS Number: 22721-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR