2-Bromo-6-methoxy-3-nitrophenol 95%

Supplier: Ambeed
Danger

A897111-100MG A897111-1G
76993-814EA 27 USD
76993-814 76993-816
2-Bromo-6-methoxy-3-nitrophenol 95%
2-Bromo-6-methoxy-3-nitrophenol

Precaución: for Research Use Only

Formula: C₇H₆BrNO₄
MDL Number: MFCD20486098
CAS Number: 854733-39-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR