4-Fluoro-3-methylbenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: (4-Fluoro-3-methylphenyl)methanamine

A896933-10G A896933-1G A896933-25G A896933-5G
77006-994EA 48.81 USD
77006-994 77006-996 77006-998 77007-000
4-Fluoro-3-methylbenzylamine 98%
4-Fluoro-3-methylbenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD01631490
CAS Number: 261951-68-2
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR