1-Bromo-2-methyl-4-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-(trifluoromethoxy)toluene

A896477-1G A896477-250MG A896477-5G
77036-828EA 38.2 USD
77036-828 77036-830 77036-832
1-Bromo-2-methyl-4-(trifluoromethoxy)benzene 98%
1-Bromo-2-methyl-4-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O
MW: 255.0319 g/mol
MDL Number: MFCD01631545
CAS Number: 261951-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR