2-(Trifluoromethyl)benzo[d]thiazol-5-amine 97%

Supplier: Ambeed
Warning

A896349-1G A896349-250MG A896349-100MG
77019-890EA 418 USD
77019-890 77019-892 77891-666
2-(Trifluoromethyl)benzo[d]thiazol-5-amine 97%
2-(Trifluoromethyl)benzo[d]thiazol-5-amine

Precaución: for Research Use Only

Formula: C₈H₅F₃N₂S
MDL Number: MFCD18821454
CAS Number: 58460-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR