3-Azabicyclo[3.1.0]hexane-2,4-dione 95%

Supplier: Ambeed
Warning

A895129-100MG A895129-250MG
76993-810EA 57.29 USD
76993-810 76993-812
3-Azabicyclo[3.1.0]hexane-2,4-dione 95%
3-Azabicyclo[3.1.0]hexane-2,4-dione

Precaución: for Research Use Only

Formula: C₅H₅NO₂
MW: 111.09 g/mol
MDL Number: MFCD00187526
CAS Number: 5617-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR