3-Cyclohexene-1-carboxylic acid 98%
Supplier: Ambeed
|
Danger
|
A894968-10G
A894968-25G
A894968-100G
77006-990EA
5
USD
77006-990
77006-992
77674-326
3-Cyclohexene-1-carboxylic acid 98%
3-Cyclohexene-1-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₁₀O₂ |
MDL Number:
MFCD00013781 CAS Number: 4771-80-6 UN: 3265 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...