3-Bromo-5-methyl-1,2,4-oxadiazole 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A893106-1G A893106-250MG A893106-100MG A893106-5G
76993-802EA 144 USD
76993-802 76993-804 77684-854 77891-664
3-Bromo-5-methyl-1,2,4-oxadiazole 98%, Ambeed.Inc
3-Bromo-5-methyl-1,2,4-oxadiazole

Precaución: For research use only.

Formula: C₃H₃BrN₂O
MDL Number: MFCD18381907
CAS Number: 1228427-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR