5-Amino-1-ethyl-1H-indazole 98%

Supplier: Ambeed
Warning

A891663-100MG A891663-1G A891663-250MG
77019-878EA 13 USD
77019-878 77019-880 77019-882
5-Amino-1-ethyl-1H-indazole 98%
5-Amino-1-ethyl-1H-indazole

Precaución: for Research Use Only

Formula: C₉H₁₁N₃
MDL Number: MFCD16620004
CAS Number: 511249-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR