Methyl-8-bromoimidazo[1,2-a]pyridine-6-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: Methyl 8-bromoimidazo[1,2-a]pyridine-6-carboxylate

A891401-1G A891401-250MG A891401-5G
76993-794EA 75.33 USD
76993-794 76993-796 76993-798
Methyl-8-bromoimidazo[1,2-a]pyridine-6-carboxylate 96%
Methyl 8-bromoimidazo[1,2-a]pyridine-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
MW: 255.0681 g/mol
MDL Number: MFCD17016026
CAS Number: 1234616-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR