Fmoc-D-isoleucine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-D-Ile-OH, (2R,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylpentanoic acid

A891101-5G A891101-1G A891101-100MG
77006-980EA 651.4 USD
77006-980 77569-620 77603-228
Fmoc-D-isoleucine 98%
Fmoc-D-isoleucine

Precaución: For research use only.

Formula: C₂₁H₂₃NO₄
MW: 353.418 g/mol
MDL Number: MFCD00080291
CAS Number: 143688-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR