Methyl 2-amino-4,5-dimethylbenzoate 98%

Supplier: Ambeed
Warning

A890498-1G A890498-250MG A890498-5G
77019-872EA 29 USD
77019-872 77019-874 77019-876
Methyl 2-amino-4,5-dimethylbenzoate 98%
Methyl 2-amino-4,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MDL Number: MFCD11505980
CAS Number: 19258-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR