4-Azabenzimidazole 97%

Supplier: Ambeed
Warning

Synonyms: 4H-Imidazo[4,5-b]pyridine

A890252-1G A890252-250MG A890252-5G
77006-974EA 54 USD
77006-974 77006-976 77006-978
4-Azabenzimidazole 97%
4-Azabenzimidazole

Precaución: for Research Use Only

Formula: C₆H₅N₃
MDL Number: MFCD00005579
CAS Number: 273-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR