tert-Butyl 8-oxo-2-azaspiro[4.5]decane-2-carboxylate 98%

Supplier: Ambeed
Warning

A889743-100MG A889743-1G A889743-250MG
77019-866EA 94.43 USD
77019-866 77019-868 77019-870
tert-Butyl 8-oxo-2-azaspiro[4.5]decane-2-carboxylate 98%
tert-Butyl 8-oxo-2-azaspiro[4.5]decane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₃NO₃
MW: 253.33 g/mol
MDL Number: MFCD18711592
CAS Number: 1272758-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR