2-(1H-Benzo[d][1,2,3]triazol-1-yl)ethanamine 97%

Supplier: Ambeed
Danger

A888697-1G A888697-250MG
76993-790EA 353.28 USD
76993-790 76993-792
2-(1H-Benzo[d][1,2,3]triazol-1-yl)ethanamine 97%
2-(1H-Benzo[d][1,2,3]triazol-1-yl)ethanamine

Precaución: for Research Use Only

Formula: C₈H₁₀N₄
MW: 162.19 g/mol
MDL Number: MFCD06213713
CAS Number: 26861-65-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR