2-(4-(tert-Butoxycarbonyl)piperazin-2-yl)acetic acid 98%

Supplier: Ambeed
Warning

A888646-100MG A888646-1G A888646-250MG
77019-860EA 79.57 USD
77019-860 77019-862 77019-864
2-(4-(tert-Butoxycarbonyl)piperazin-2-yl)acetic acid 98%
2-(4-(tert-Butoxycarbonyl)piperazin-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₄
MW: 244.28 g/mol
MDL Number: MFCD03001351
CAS Number: 183591-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR