2-(5-Hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

Synonyms: 5-Hexylthiopheneboronic acid pinacol ester

A888432-1G A888432-5G
76993-786EA 59.42 USD
76993-786 76993-788
2-(5-Hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(5-Hexylthiophen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₂₇BO₂S
MW: 294.26 g/mol
MDL Number: MFCD10567052
CAS Number: 917985-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR