2-(2-(2-(2-Hydroxyethoxy)ethoxy)ethoxy)ethyl 4-methylbenzenesulfonate 98%

Supplier: Ambeed
Danger

Synonyms: Tetraethylene glycol monotosylate

A887981-1G A887981-250MG
77019-858EA 84.88 USD
77019-858 77684-847
2-(2-(2-(2-Hydroxyethoxy)ethoxy)ethoxy)ethyl 4-methylbenzenesulfonate 98%
2-(2-(2-(2-Hydroxyethoxy)ethoxy)ethoxy)ethyl 4-methylbenzenesulfonate

Precaución: For research use only.

Formula: C₁₅H₂₄O₇S
MW: 348.41 g/mol
MDL Number: MFCD22056303
CAS Number: 77544-60-6
UN: 2927
ADR: 6.1,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR