cis-4-Aminocyclohexanol 98%

Supplier: Ambeed
Danger

A887798-250MG A887798-5G A887798-1G
77036-812EA 14 USD
77036-812 77552-670 77569-614
cis-4-Aminocyclohexanol 98%
cis-4-Aminocyclohexanol

Precaución: For research use only.

Formula: C₆H₁₃NO
MDL Number: MFCD09997786
CAS Number: 40525-78-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR