2,6-Diazaspiro[3.3]heptane dihydrochloride 98%

Supplier: Ambeed
Warning

A887068-100MG A887068-250MG
76993-782EA 122 USD
76993-782 76993-784
2,6-Diazaspiro[3.3]heptane dihydrochloride 98%
2,6-Diazaspiro[3.3]heptane dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂Cl₂N₂
MW: 171.06 g/mol
MDL Number: MFCD22576062
CAS Number: 1184963-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR