8-Methylene-1,4-dioxa-spiro[4.5]decane 95% stabilized

Supplier: Ambeed
Warning

Synonyms: 8-Methylene-1,4-dioxaspiro[4.5]decane

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A886444-1G A886444-250MG A886444-5G
77036-806EA 196.27 USD
77036-806 77036-808 77036-810
8-Methylene-1,4-dioxa-spiro[4.5]decane 95% stabilized
8-Methylene-1,4-dioxa-spiro[4.5]decane

Precaución: for Research Use Only

Formula: C₉H₁₄O₂
MW: 154.206 g/mol
MDL Number: MFCD07779440
CAS Number: 51656-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR