N-Ethylpyridin-3-amine 98%

Supplier: Ambeed
Danger

A886324-1G A886324-250MG A886324-5G
76993-776EA 44.56 USD
76993-776 76993-778 76993-780
N-Ethylpyridin-3-amine 98%
N-Ethylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.168 g/mol
MDL Number: MFCD11140216
CAS Number: 32405-70-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR