H-D-Phe(4-F)-OH·HCl 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Amino-3-(4-fluorophenyl)propanoic acid hydrochloride

A885494-1G A885494-250MG A885494-25G A885494-5G
77006-960EA 29.71 USD
77006-960 77006-962 77006-964 77006-966
H-D-Phe(4-F)-OH·HCl 98%
H-D-Phe(4-F)-OH·HCl

Precaución: for Research Use Only

Formula: C₉H₁₀NO₂F·HCl
MW: 219.7 g/mol
MDL Number: MFCD01321254
CAS Number: 122839-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR