1-Phenyl-5-pyrazolone-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Oxo-1-phenyl-4,5-dihydro-1H-pyrazole-3-carboxylic acid, 5-Oxo-1-phenyl-2-pyrazolin-3-carboxylic Acid

A885110-25G A885110-5G
77006-956EA 24.41 USD
77006-956 77006-958
1-Phenyl-5-pyrazolone-3-carboxylic acid 98%
1-Phenyl-5-pyrazolone-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₃
MW: 204.19 g/mol
MDL Number: MFCD00037318
CAS Number: 119-18-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR