3-Bromo-5-(hydroxymethyl)benzonitrile 95%

Supplier: Ambeed
Warning

A884921-100MG A884921-1G A884921-250MG
76993-766EA 90 USD
76993-766 76993-768 76993-770
3-Bromo-5-(hydroxymethyl)benzonitrile 95%
3-Bromo-5-(hydroxymethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MDL Number: MFCD09261039
CAS Number: 1205515-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR