N1,N1-Dibutyl-N4,N4-bis(4-(dibutylamino)phenyl)benzene-1,4-diamine 95%

Supplier: Ambeed
Warning

A884568-1G A884568-250MG A884568-5G
76993-760EA 95 USD
76993-760 76993-762 76993-764
N1,N1-Dibutyl-N4,N4-bis(4-(dibutylamino)phenyl)benzene-1,4-diamine 95%
N1,N1-Dibutyl-N4,N4-bis(4-(dibutylamino)phenyl)benzene-1,4-diamine

Precaución: for Research Use Only

Formula: C₄₂H₆₆N₄
MW: 627.00 g/mol
MDL Number: MFCD22056142
CAS Number: 47862-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR